Stampa…

5-Bromo-2-methylpyrimidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5266-1G B5266-5G
TCIAB5266-1GEA 28 EUR
TCIAB5266-1G TCIAB5266-5G
5-Bromo-2-methylpyrimidine ≥98.0% (by GC)
5-Bromo-2-methylpyrimidine
Formula: C₅H₅BrN₂
Peso molecolare: 173,01 g/mol
MDL Number: MFCD07375143
CAS Number: 7752-78-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=effcd7570de870d2371a310e9cf1a76e01f92b6b&vid=effcd7570de870d2371a310e9cf1a76e01f92b6b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 85.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR