Bis(2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₉H₂₂F₃IO₃S |
MDL Number:
MFCD25541454 CAS Number: 139139-80-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...