4-Benzyloxy-2-formylphenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(4-(Benzyloxy)-2-formylphenyl)boronic acid
B5257-200MG
B5257-1G
TCIAB5257-200MGEA
29
EUR
TCIAB5257-200MG
TCIAB5257-1G
4-Benzyloxy-2-formylphenylboronic acid (contains varying amounts of Anhydride)
4-Benzyloxy-2-formylphenylboronic acid
Formula:
C₁₄H₁₃BO₄ Peso molecolare: 256,0616 g/mol |
MDL Number:
MFCD02179486 CAS Number: 139962-97-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 98.0 to 107.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...