Stampa…

6,7-Bis(benzyloxy)coumarin ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

B5251-200MG B5251-1G
TCIAB5251-200MGEA 47 EUR
TCIAB5251-200MG TCIAB5251-1G
6,7-Bis(benzyloxy)coumarin ≥98.0% (by HPLC)
6,7-Bis(benzyloxy)coumarin
Formula: C₂₃H₁₈O₄
Peso molecolare: 358,39 g/mol
MDL Number: MFCD00016975
CAS Number: 909-84-2
EINECS: 213-003-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 161.0 to 165.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR