3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
Synonyms:
1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxamide, 1-Boc-3-pyrrolidinecarboxamide, tert-Butyl 3-Carbamoylpyrrolidine-1-carboxylate, 3-Carbamoylpyrrolidine-1-carboxylic acid tert-Butyl ester, 1-Boc-3-Carbamoylpyrrolidine
B5248-1G
B5248-5G
TCIAB5248-1GEA
107
EUR
TCIAB5248-1G
TCIAB5248-5G
3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester ≥98.0% (by HPLC, total nitrogen)
3-Carbamoylpyrrolidine-1-carboxylic acid tert-butyl ester
Formula:
C₁₀H₁₈N₂O₃ Peso molecolare: 214,27 g/mol |
MDL Number:
MFCD04116210 CAS Number: 122684-34-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...