Stampa…

N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulphonyloxy)-L-proline methyl ester ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Boc-trans-4-Tosyloxy-L-proline methyl ester, Boc-Hyp(Tos)-OMe, (2S,4R)-1-tert-Butyl 2-Methyl 4-(Tosyloxy)pyrrolidine-1,2-dicarboxylate, N-Boc-trans-4-(p-tosyloxy)-L-proline methyl ester, (4R)-4-TsO-N-Boc-Pro-OMe

B5247-200MG B5247-1G
TCIAB5247-200MGEA 22 EUR
TCIAB5247-200MG TCIAB5247-1G
N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulphonyloxy)-L-proline methyl ester ≥98.0% (by HPLC, total nitrogen)
N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulphonyloxy)-L-proline methyl ester
Formula: C₁₈H₂₅NO₇S
Peso molecolare: 399,46 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD02179383
CAS Number: 88043-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=519f91c5756564f4a4cc1a5c70de8b40b544169b&vid=519f91c5756564f4a4cc1a5c70de8b40b544169b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 77.0 to 81.0 deg-C
Specific rotation [a]20/D -33.0 to -38.0 deg (C=0.7, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR