Stampa…

4-Benzyloxyaniline ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(Benzyloxy)aniline

B5227-5G B5227-25G
TCIAB5227-5GEA 60 EUR
TCIAB5227-5G TCIAB5227-25G
4-Benzyloxyaniline ≥98.0% (by GC, titration analysis)
4-Benzyloxyaniline
Formula: C₁₃H₁₃NO
Peso molecolare: 199,2484 g/mol
Punto di ebollizione: 155°C
MDL Number: MFCD00025318
CAS Number: 6373-46-2
EINECS: 228-917-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR