Stampa…

1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine ≥95.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: tert-Butyl 3-(Cyanomethylene)azetidine-1-carboxylate, 3-(Cyanomethylene)azetidine-1-carboxylic acid tert-Butyl ester, 1-Boc-3-(cyanomethylene)azetidine

B5217-200MG B5217-1G
TCIAB5217-200MGEA 27 EUR
TCIAB5217-200MG TCIAB5217-1G
1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine ≥95.0% (by GC)
1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine
Formula: C₁₀H₁₄N₂O₂
Peso molecolare: 194,23 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD12761241
CAS Number: 1153949-11-1
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b928649175ec8cd2e19d3d0638a1ca735a67b15&vid=4b928649175ec8cd2e19d3d0638a1ca735a67b15&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR