Stampa…

5-Bromo-6-hydroxynicotinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B5193-1G B5193-5G
TCIAB5193-1GEA 44 EUR
TCIAB5193-1G TCIAB5193-5G
5-Bromo-6-hydroxynicotinic acid ≥98.0% (by GC, titration analysis)
5-Bromo-6-hydroxynicotinic acid
Formula: C₆H₄BrNO₃
Punto di fusione: 296°C
MDL Number: MFCD08235173
CAS Number: 41668-13-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdd48c2c755e9e787dd9ad4d8cf39d6e50b5ea8b&vid=cdd48c2c755e9e787dd9ad4d8cf39d6e50b5ea8b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR