Stampa…

Butylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Butylamine Hydrobromide, BABr, Butylammonium bromide

B5186-1G B5186-5G
TCIAB5186-1GEA 27 EUR
TCIAB5186-1G TCIAB5186-5G
Butylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Butylamine hydrobromide
Formula: C₄H₁₁N·HBr
Peso molecolare: 154,05 g/mol
MDL Number: MFCD01874353
CAS Number: 15567-09-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR