Benzylamine hydrobromide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Benzylammonium bromide
Formula:
C₇H₉N·HBr Punto di fusione: 204°C |
MDL Number:
MFCD00035434 CAS Number: 37488-40-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...