Stampa…

3-Bromoanthranilic acid ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-3-bromobenzoic acid

B5177-1G B5177-5G
TCIAB5177-1GEA 34 EUR
TCIAB5177-1G TCIAB5177-5G
3-Bromoanthranilic acid ≥98.0% (by HPLC)
3-Bromoanthranilic acid
Formula: C₇H₆BrNO₂
Peso molecolare: 216,03 g/mol
MDL Number: MFCD03618453
CAS Number: 20776-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de2c36275402346cac32053cad55bf33f25163ab&vid=de2c36275402346cac32053cad55bf33f25163ab&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 175.0 to 179.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR