6-Bromo-4-chloroquinoline ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B5166-1G
B5166-5G
TCIAB5166-1GEA
63
EUR
TCIAB5166-1G
TCIAB5166-5G
6-Bromo-4-chloroquinoline ≥98.0% (by GC)
6-Bromo-4-chloroquinoline
Formula:
C₉H₅BrClN Peso molecolare: 242,50 g/mol |
MDL Number:
MFCD00511001 CAS Number: 65340-70-7 |
Risultati del test di specificazione
Purity (GC) | min. 98.0 % |
Melting point | 111.0 to 115.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...