Stampa…

(4-Bromo-2-nitrophenyl)acetic acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Bromo-2-nitrophenylacetic acid

B5159-1G B5159-5G
TCIAB5159-1GEA 69 EUR
TCIAB5159-1G TCIAB5159-5G
(4-Bromo-2-nitrophenyl)acetic acid ≥97.0% (by titrimetric analysis)
(4-Bromo-2-nitrophenyl)acetic acid
Formula: C₈H₆BrNO₄
Peso molecolare: 260,0400 g/mol
MDL Number: MFCD05663950
CAS Number: 6127-11-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edd13bfc305e4ac7b1adc54664e6e56aafe8ba07&vid=edd13bfc305e4ac7b1adc54664e6e56aafe8ba07&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 165.0 to 169.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR