[(4-Bromobenzyl)oxy](tert-butyl)dimethylsilane ≥98.0% (by GC)
Fornitore: TCI
B5155-5G
B5155-25G
TCIAB5155-5GEA
97
EUR
TCIAB5155-5G
TCIAB5155-25G
[(4-Bromobenzyl)oxy](tert-butyl)dimethylsilane ≥98.0% (by GC)
[(4-Bromobenzyl)oxy](tert-butyl)dimethylsilane
Formula:
C₁₃H₂₁BrOSi Punto di ebollizione: 160°C |
MDL Number:
MFCD07368899 CAS Number: 87736-74-1 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...