9,10-Bis[N-(p-tolyl)anilino]anthracene ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
Synonyms:
N,N'-Bis(4-methylphenyl)-N,N'-diphenyl-9,10-anthracenediamine, N,N'-Diphenyl-N,N'-di(p-tolyl)-9,10-anthracenediamine
B5149-200MG
B5149-1G
TCIAB5149-200MGEA
70
EUR
TCIAB5149-200MG
TCIAB5149-1G
9,10-Bis[N-(p-tolyl)anilino]anthracene ≥98.0% (by HPLC, total nitrogen)
9,10-Bis[N-(p-tolyl)anilino]anthracene
Formula:
C₄₀H₃₂N₂ Peso molecolare: 540,71 g/mol |
MDL Number:
MFCD12022466 CAS Number: 190974-21-1 |
Risultati del test di specificazione
Appearance | Yellow to Amber powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...