Stampa…

trans-1,4-Bis(aminomethyl)cyclohexane ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: trans-1,4-Di(aminomethyl)cyclohexane, trans-Hexahydro-p-xylylenediamine

B5147-1G B5147-5G
TCIAB5147-1GEA 235 EUR
TCIAB5147-1G TCIAB5147-5G
trans-1,4-Bis(aminomethyl)cyclohexane ≥98.0% (by GC, titration analysis)
trans-1,4-Bis(aminomethyl)cyclohexane
Formula: C₈H₁₈N₂
Peso molecolare: 142,246 g/mol
Punto di ebollizione: 118°C
MDL Number: MFCD20037427
CAS Number: 10029-07-9
UN: 2922
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR