Stampa…

4,4'-Bis(2,2-diphenylvinyl)biphenyl ≥99.0% (by HPLC), purified by sublimation

Fornitore: TCI

B5139-200MG B5139-1G
TCIAB5139-200MGEA 36 EUR
TCIAB5139-200MG TCIAB5139-1G
4,4'-Bis(2,2-diphenylvinyl)biphenyl ≥99.0% (by HPLC), purified by sublimation
4,4'-Bis(2,2-diphenylvinyl)biphenyl
Formula: C₄₀H₃₀
MDL Number: MFCD03093280
CAS Number: 142289-08-5

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 99.0 area%
Melting point 207.0 to 211.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR