Stampa…

3-Bromo-6-fluoro-2-methylpyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Bromo-2-fluoro-6-methylpyridine, 5-Bromo-2-fluoro-6-picoline

B5135-5G B5135-25G
TCIAB5135-5GEA 40 EUR
TCIAB5135-5G TCIAB5135-25G
3-Bromo-6-fluoro-2-methylpyridine ≥98.0% (by GC)
3-Bromo-6-fluoro-2-methylpyridine
Formula: C₆H₅BrFN
Peso molecolare: 190,01 g/mol
MDL Number: MFCD03095092
CAS Number: 375368-83-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=632c7d8df4c6fd2da4d2af71451f68382bc82e57&vid=632c7d8df4c6fd2da4d2af71451f68382bc82e57&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR