2-Bromo-5-chloro-3-methylpyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Bromo-5-chloro-3-picoline
B5133-5G
B5133-25G
TCIAB5133-5GEA
46
EUR
TCIAB5133-5G
TCIAB5133-25G
2-Bromo-5-chloro-3-methylpyridine ≥98.0% (by GC)
2-Bromo-5-chloro-3-methylpyridine
Formula:
C₆H₅BrClN Peso molecolare: 206,47 g/mol |
MDL Number:
MFCD03095062 CAS Number: 65550-77-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...