Stampa…

3-Bromo-2-methoxyaniline ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: 3-Bromo-o-anisidine, 2-Amino-6-bromoanisole

B5131-1G B5131-5G
TCIAB5131-1GEA 75 EUR
TCIAB5131-1G TCIAB5131-5G
3-Bromo-2-methoxyaniline ≥98.0% (by GC, titration analysis)
3-Bromo-2-methoxyaniline
Formula: C₇H₈BrNO
Punto di ebollizione: 159°C
MDL Number: MFCD13186737
CAS Number: 116557-46-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c52483f4b0b8caf829771c32bbb602a8590db88d&vid=c52483f4b0b8caf829771c32bbb602a8590db88d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR