9-(4-Biphenylyl)carbazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B5128-1G
B5128-5G
TCIAB5128-1GEA
37
EUR
TCIAB5128-1G
TCIAB5128-5G
9-(4-Biphenylyl)carbazole ≥98.0% (by GC)
9-(4-Biphenylyl)carbazole
Formula:
C₂₄H₁₇N Peso molecolare: 319,41 g/mol |
MDL Number:
MFCD28147704 CAS Number: 6299-16-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 225.0 to 229.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...