Stampa…

trans,trans-4-Butyl-4'-vinylbicyclohexyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

B5123-1G B5123-5G
TCIAB5123-1GEA 31 EUR
TCIAB5123-1G TCIAB5123-5G
trans,trans-4-Butyl-4'-vinylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-Butyl-4'-vinylbicyclohexyl
Formula: C₁₈H₃₂
Peso molecolare: 248,45 g/mol
MDL Number: MFCD25541699
CAS Number: 153429-47-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3114eabcdbfbc9697dd1943fff5e8b0ca70f6939&vid=3114eabcdbfbc9697dd1943fff5e8b0ca70f6939&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.000 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR