4-(Benzyloxy)benzyl chloride ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₁₄H₁₃ClO Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00000916 CAS Number: 836-42-0 EINECS: 212-648-3 UN: 3261 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...