Stampa…

2-Bromo-4-iodopyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5119-1G B5119-5G
TCIAB5119-1GEA 40 EUR
TCIAB5119-1G TCIAB5119-5G
2-Bromo-4-iodopyridine ≥98.0% (by GC)
2-Bromo-4-iodopyridine
Formula: C₅H₃BrIN
Peso molecolare: 283,89 g/mol
MDL Number: MFCD03085768
CAS Number: 100523-96-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0959290dbee4dec69079573d0ef0b2f7dc5154e9&vid=0959290dbee4dec69079573d0ef0b2f7dc5154e9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR