Stampa…

9-(4-Biphenylyl)-2-bromocarbazole ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-([1,1'-Biphenyl]-4-yl)-2-bromo-9H-carbazole

B5110-200MG B5110-1G
TCIAB5110-200MGEA 35 EUR
TCIAB5110-200MG TCIAB5110-1G
9-(4-Biphenylyl)-2-bromocarbazole ≥98.0% (by HPLC, total nitrogen)
9-(4-Biphenylyl)-2-bromocarbazole
Formula: C₂₄H₁₆BrN
Peso molecolare: 398,30 g/mol
MDL Number: MFCD27979953
CAS Number: 1393835-87-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9031f68401449f048b2e08fbb6877796c5191c3&vid=a9031f68401449f048b2e08fbb6877796c5191c3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 173.0 to 177.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR