Stampa…

9-(4'-Bromo-4-biphenylyl)carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-(4'-Bromo-[1,1'-biphenyl]-4-yl)-9H-carbazole

B5109-1G B5109-5G
TCIAB5109-1GEA 77 EUR
TCIAB5109-1G TCIAB5109-5G
9-(4'-Bromo-4-biphenylyl)carbazole ≥98.0% (by GC)
9-(4'-Bromo-4-biphenylyl)carbazole
Formula: C₂₄H₁₆BrN
Peso molecolare: 398,30 g/mol
MDL Number: MFCD27644692
CAS Number: 212385-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c18a4e7c036a4b6faaa41c2fdeb063a3547deaaa&vid=c18a4e7c036a4b6faaa41c2fdeb063a3547deaaa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR