Stampa…

2-(3-Bromophenyl)-9-phenyl-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5108-1G B5108-200MG
TCIAB5108-1GEA 150 EUR
TCIAB5108-1G TCIAB5108-200MG
2-(3-Bromophenyl)-9-phenyl-9H-carbazole ≥98.0% (by GC)
2-(3-Bromophenyl)-9-phenyl-9H-carbazole
Formula: C₂₄H₁₆BrN
Peso molecolare: 398,30 g/mol
MDL Number: MFCD28147706
CAS Number: 1365118-41-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 148.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR