Stampa…

(R)-Baclofen ≥95.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (R)-4-Amino-3-(4-chlorophenyl)butanoic acid, (R)-4-Amino-3-(4-chlorophenyl)butyric acid

B5102-25MG B5102-100MG
TCIAB5102-25MGEA 68 EUR
TCIAB5102-25MG TCIAB5102-100MG
(R)-Baclofen ≥95.0% (by HPLC, titration analysis)
(R)-Baclofen
Formula: C₁₀H₁₂ClNO₂
Peso molecolare: 213,66 g/mol
MDL Number: MFCD01321057
CAS Number: 69308-37-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15d54a249c137b9867ba809dcb24e79649046d10&vid=15d54a249c137b9867ba809dcb24e79649046d10&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D 7.0 to 8.0 deg (C=1.6mol/L HCL)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR