Stampa…

Bis(4-formylphenyl) ether ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-(4-Formylphenoxy)benzaldehyde, 4,4'-Diformyldiphenyl ether, 4,4'-Oxydibenzaldehyde

B5099-1G B5099-5G
TCIAB5099-1GEA 91 EUR
TCIAB5099-1G TCIAB5099-5G
Bis(4-formylphenyl) ether ≥98.0% (by GC)
Bis(4-formylphenyl) ether
Formula: C₁₄H₁₀O₃
Peso molecolare: 226,23 g/mol
Punto di ebollizione: 160°C
MDL Number: MFCD08276812
CAS Number: 2215-76-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41064fea16a3864aa03c969e1bdd8e8cef8b9820&vid=41064fea16a3864aa03c969e1bdd8e8cef8b9820&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR