Stampa…

4-Bromo-2,6-difluorobenzyl alcohol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (4-Bromo-2,6-difluorophenyl)methanol

B5090-1G B5090-5G
TCIAB5090-1GEA 39 EUR
TCIAB5090-1G TCIAB5090-5G
4-Bromo-2,6-difluorobenzyl alcohol ≥98.0% (by GC)
4-Bromo-2,6-difluorobenzyl alcohol
Formula: C₇H₅BrF₂O
Peso molecolare: 223,02 g/mol
MDL Number: MFCD03094461
CAS Number: 162744-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfe5e28d1e59d5405e9dec086f6ba7c90bc61d49&vid=cfe5e28d1e59d5405e9dec086f6ba7c90bc61d49&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR