5-Bromo-2-chloro-3-fluoropyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B5089-5G
B5089-25G
TCIAB5089-5GEA
49
EUR
TCIAB5089-5G
TCIAB5089-25G
5-Bromo-2-chloro-3-fluoropyridine ≥98.0% (by GC)
5-Bromo-2-chloro-3-fluoropyridine
Formula:
C₅H₂BrClFN Peso molecolare: 210,43 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD03411574 CAS Number: 831203-13-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 50.0 to 54.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...