Stampa…

N,N-Bis(4-biphenylyl)aniline ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: N-([1,1'-Biphenyl]-4-yl)-N-phenyl-[1,1'-biphenyl]-4-amine

B5086-1G B5086-5G
TCIAB5086-1GEA 73 EUR
TCIAB5086-1G TCIAB5086-5G
N,N-Bis(4-biphenylyl)aniline ≥98.0% (by HPLC, total nitrogen)
N,N-Bis(4-biphenylyl)aniline
Formula: C₃₀H₂₃N
Peso molecolare: 397,52 g/mol
MDL Number: MFCD20483034
CAS Number: 122215-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db41a053839b6ee653fd823c65cb41fc1bdcff59&vid=db41a053839b6ee653fd823c65cb41fc1bdcff59&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 169.0 to 173.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR