Stampa…

1,2-Bis(4-methoxyphenyl)-2-oxoethyl Cyclohexylcarbamate ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: Cyclohexylcarbamic acid 1,2-Bis(4-methoxyphenyl)-2-oxoethyl ester, 1,2-Bis(4-methoxyphenyl)-2-oxoethyl cyclohexylcarbamate

B5085-200MG B5085-1G
TCIAB5085-200MGEA 65 EUR
TCIAB5085-200MG TCIAB5085-1G
1,2-Bis(4-methoxyphenyl)-2-oxoethyl Cyclohexylcarbamate ≥98.0% (by HPLC, total nitrogen)
1,2-Bis(4-methoxyphenyl)-2-oxoethyl Cyclohexylcarbamate
Formula: C₂₃H₂₇NO₅
Peso molecolare: 397,47 g/mol
MDL Number: MFCD29472516
CAS Number: 196599-80-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR