Borane - Ammonia Complex ≥85.0% (V)
Fornitore: TCI EUROPE
Synonyms:
Ammonia Borane
B5082-5G
B5082-25G
TCIAB5082-5GEA
100
EUR
TCIAB5082-5G
TCIAB5082-25G
Borane - Ammonia Complex ≥85.0% (V)
Borane - Ammonia Complex
Formula:
NH₃·BH₃ Peso molecolare: 30,87 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00075639 CAS Number: 13774-81-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Gasburet method) | min. 85.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...