6-Bromo-2-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Bromo-2-pyridylboronic acid Pinacol ester, 6-Bromopyridine-2-boronic acid pinacol ester, 2-(6-Bromo-2-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B5077-1G
B5077-5G
TCIAB5077-1GEA
54
EUR
TCIAB5077-1G
TCIAB5077-5G
6-Bromo-2-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
6-Bromo-2-pyridineboronic acid pinacol ester
Formula:
C₁₁H₁₅BBrNO₂ Peso molecolare: 283,96 g/mol |
MDL Number:
MFCD06796177 CAS Number: 651358-83-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...