Stampa…

4-Bromo-2-fluoro-N-methylbenzamide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

B5074-1G B5074-5G
TCIAB5074-1GEA 33 EUR
TCIAB5074-1G TCIAB5074-5G
4-Bromo-2-fluoro-N-methylbenzamide ≥98.0% (by HPLC, total nitrogen)
4-Bromo-2-fluoro-N-methylbenzamide
Formula: C₈H₇BrFNO
Peso molecolare: 232,0497 g/mol
MDL Number: MFCD09878362
CAS Number: 749927-69-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 125.0 to 129.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR