1,3-Benzenediboronic acid bis(pinacol) ester ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
1,3-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 1,3-Phenylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
Formula:
C₁₈H₂₈B₂O₄ |
MDL Number:
MFCD09864185 CAS Number: 196212-27-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 133.0 to 137.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...