Stampa…

1,2-Benzenediboronic acid bis(pinacol) ester ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,2-Phenylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 2,2-(1,2-Phenylene)bis[4,4,5,5-tetramethyl-1,3,2-dioxaborolane], 1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

B5069-1G B5069-5G
TCIAB5069-1GEA 113 EUR
TCIAB5069-1G TCIAB5069-5G
1,2-Benzenediboronic acid bis(pinacol) ester ≥98.0% (by GC)
1,2-Benzenediboronic acid bis(pinacol) ester
Formula: C₁₈H₂₈B₂O₄
Peso molecolare: 330,04 g/mol
MDL Number: MFCD28969469
CAS Number: 269410-07-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR