2-(5-Bromo-1H-indol-3-yl)-1-ethanol ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
5-Bromoindole-3-ethanol, 2-(5-Bromo-1H-indol-3-yl)ethanol, 5-Bromo-3-(2-hydroxyethyl)indole
B5068-200MG
B5068-1G
TCIAB5068-200MGEA
42
EUR
TCIAB5068-200MG
TCIAB5068-1G
2-(5-Bromo-1H-indol-3-yl)-1-ethanol ≥98.0% (by GC)
2-(5-Bromo-1H-indol-3-yl)-1-ethanol
Formula:
C₁₀H₁₀BrNO Peso molecolare: 240,11 g/mol |
MDL Number:
MFCD00130167 CAS Number: 32774-29-1 EINECS: 251-205-9 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...