Stampa…

Bisacodyl ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

B5066-5G B5066-25G
TCIAB5066-5GEA 47 EUR
TCIAB5066-5G TCIAB5066-25G
Bisacodyl ≥98.0% (by HPLC, titration analysis)
Bisacodyl
Formula: C₂₂H₁₉NO₄
MDL Number: MFCD00038039
CAS Number: 603-50-9
EINECS: 210-044-4
Merck Index: 14,01242
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f355cea94c8543e9fbd96340b27860e2c8929df&vid=3f355cea94c8543e9fbd96340b27860e2c8929df&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR