Stampa…

5-Bromo-2-fluorobenzeneboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

Synonyms: 5-Bromo-2-fluorophenylboronic acid, 5-Bromo-2-fluorobenzeneboronic acid

B5065-25G B5065-5G
TCIAB5065-25GEA 267 EUR
TCIAB5065-25G TCIAB5065-5G
5-Bromo-2-fluorobenzeneboronic acid (contains varying amounts of Anhydride)
5-Bromo-2-fluorobenzeneboronic acid
Formula: C₆H₅BBrFO₂
Punto di fusione: 174°C
MDL Number: MFCD05664307
CAS Number: 112204-57-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR