Stampa…

3-Bromo-2-chloro-5-methylpyridine ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Bromo-2-chloro-5-picoline

B5061-5G B5061-25G
TCIAB5061-5GEA 80 EUR
TCIAB5061-5G TCIAB5061-25G
3-Bromo-2-chloro-5-methylpyridine ≥96.0% (by GC)
3-Bromo-2-chloro-5-methylpyridine
Formula: C₆H₅BrClN
MDL Number: MFCD01830664
CAS Number: 17282-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b1978cb0ee3e07ad1843fcb3f1ef6a118b69f93&vid=1b1978cb0ee3e07ad1843fcb3f1ef6a118b69f93&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR