Stampa…

3-(tert-Butyl)-9H-carbazole ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5053-200MG B5053-1G
TCIAB5053-200MGEA 59 EUR
TCIAB5053-200MG TCIAB5053-1G
3-(tert-Butyl)-9H-carbazole ≥95.0% (by GC)
3-(tert-Butyl)-9H-carbazole
Formula: C₁₆H₁₇N
Peso molecolare: 223,32 g/mol
MDL Number: MFCD12024281
CAS Number: 22401-74-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=890156eaa9a14e203580264af827636566ba6a82&vid=890156eaa9a14e203580264af827636566ba6a82&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 151.0 to 155.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR