Stampa…

N-(4'-Bromo-4-biphenylyl)-N-phenyl-1-naphthylamine ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: N-(4'-Bromo-[1,1'-biphenyl]-4-yl)-N-phenylnaphthalen-1-amine

B5049-200MG B5049-1G
TCIAB5049-200MGEA 37 EUR
TCIAB5049-200MG TCIAB5049-1G
N-(4'-Bromo-4-biphenylyl)-N-phenyl-1-naphthylamine ≥98.0% (by GC)
N-(4'-Bromo-4-biphenylyl)-N-phenyl-1-naphthylamine
Formula: C₂₈H₂₀BrN
Peso molecolare: 450,38 g/mol
MDL Number: MFCD24387457
CAS Number: 352359-42-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 167.0 to 171.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR