Stampa…

9,10-Bis(6-methoxy-2-naphthyl)anthracene ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

B5046-200MG B5046-1G
TCIAB5046-200MGEA 118 EUR
TCIAB5046-200MG TCIAB5046-1G
9,10-Bis(6-methoxy-2-naphthyl)anthracene ≥98.0% (by HPLC)
9,10-Bis(6-methoxy-2-naphthyl)anthracene
Formula: C₃₆H₂₆O₂
Peso molecolare: 490,60 g/mol
MDL Number: MFCD12022382
CAS Number: 235099-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adb74869de1283bc1de610408c5a26d52030135d&vid=adb74869de1283bc1de610408c5a26d52030135d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR