Stampa…

N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE

Synonyms: N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-dimethyl-N-phenyl-9H-fluoren-2-amine

B5045-200MG B5045-1G
TCIAB5045-200MGEA 43 EUR
TCIAB5045-200MG TCIAB5045-1G
N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline ≥98.0% (by total nitrogen basis)
N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline
Formula: C₃₆H₃₁N
Peso molecolare: 477,65 g/mol
MDL Number: MFCD29089346
CAS Number: 165320-27-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR