3-Bromo-9-(2-naphthyl)carbazole ≥97.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Bromo-9-(naphthalen-2-yl)-9H-carbazole
B5028-1G
B5028-5G
TCIAB5028-1GEA
47
EUR
TCIAB5028-1G
TCIAB5028-5G
3-Bromo-9-(2-naphthyl)carbazole ≥97.0% (by HPLC, total nitrogen)
3-Bromo-9-(2-naphthyl)carbazole
Formula:
C₂₂H₁₄BrN Peso molecolare: 372,27 g/mol |
MDL Number:
MFCD26127422 CAS Number: 934545-80-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...