Stampa…

9-(3-Biphenylyl)-3-bromocarbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-([1,1'-Biphenyl]-3-yl)-3-bromo-9H-carbazole

B5024-1G B5024-5G
TCIAB5024-1GEA 79 EUR
TCIAB5024-1G TCIAB5024-5G
9-(3-Biphenylyl)-3-bromocarbazole ≥98.0% (by GC)
9-(3-Biphenylyl)-3-bromocarbazole
Formula: C₂₄H₁₆BrN
Peso molecolare: 398,30 g/mol
MDL Number: MFCD28138081
CAS Number: 1428551-28-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 112.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR