Stampa…

(4-Bromobutyl)phosphonic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

B5014-200MG B5014-1G
TCIAB5014-200MGEA 114 EUR
TCIAB5014-200MG TCIAB5014-1G
(4-Bromobutyl)phosphonic acid ≥98.0% (by titrimetric analysis)
(4-Bromobutyl)phosphonic acid
Formula: C₄H₁₀BrO₃P
Peso molecolare: 217,00 g/mol
MDL Number: MFCD00466589
CAS Number: 1190-14-3
UN: 3261
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 127.0 to 131.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR